Assay Details
| Gene Symbol: Clec4a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005860.2 | NP_001005860.1 | 5 - 6 | 566 | 152 | |
| GenBank mRNA | AY397673.1 | - | 5 - 6 | 566 | 152 | |
| BC104370.1 | - | 5 - 6 | 566 | 40049888 | 152 | |
| BC104371.2 | - | 5 - 6 | 565 | 152 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103887 | BX524711.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103887 | NM_001005860.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103887 | AY397673.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 448856 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 557597 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588392 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 708390 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389544 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 427325 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555984 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841273 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 812774 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555203 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 899040 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55183 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76378 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 166245 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103887 | BX524711.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103887 | NM_001005860.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103887 | AY397673.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24811287 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01436467 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06053507 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01436463 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06053497 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06053497 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01436467 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05352365 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05352375 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05352366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24811287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24811256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05462614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24811281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05462610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24811277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24811257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24811283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05352368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24811266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24811272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24811259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24811264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24811275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24811270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24811287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24811289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24811258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24811280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24811278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05462602 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10541578 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10541577 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10541580 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10541576 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10541579 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10541575 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 922417 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161114 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161114 | - | - |
| mouse-6 v1.1 (Illumina) | GI_54312069-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3161114 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161114 | - | - |

Clec4a4
Genomic Map