| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100014591.mm.1 |
NM_001007461.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100014591.mm.1 |
NM_001007461.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100014591.mm.1 |
TC1100001575.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100014587.mm.1 |
NM_001007461.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100014587.mm.1 |
NM_001007461.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100014587.mm.1 |
TC1100001575.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100014589.mm.1 |
NM_001007461.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100014589.mm.1 |
NM_001007461.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100014589.mm.1 |
TC1100001575.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100014592.mm.1 |
NM_001007461.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100014592.mm.1 |
NM_001007461.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100014592.mm.1 |
TC1100001575.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100014590.mm.1 |
NM_001007461.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100014590.mm.1 |
NM_001007461.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100014590.mm.1 |
TC1100001575.mm.1 |
MTA TC |