Assay Details
| Gene Symbol: Gsdma3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007461.1 | NP_001007462.1 | 5 - 6 | 737 | 135 | |
| GenBank mRNA | AY679090.1 | - | 5 - 6 | 737 | 135 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gsdma3
Genomic Map