Assay Details
| Gene Symbol: Gsdma3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007461.1 | NP_001007462.1 | 8 - 9 | 1020 | 137 | |
| GenBank mRNA | AY679090.1 | - | 8 - 9 | 1020 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
hair follicle development
sebaceous gland cell differentiation
multicellular organism development
hair follicle morphogenesis
somatic stem cell population maintenance
avascular cornea development in camera-type eye
hair cycle
catagen
anagen
positive regulation of apoptotic process
skin development
positive regulation of extrinsic apoptotic signaling pathway
molecular_function
hair follicle development
sebaceous gland cell differentiation
multicellular organism development
hair follicle morphogenesis
somatic stem cell population maintenance
avascular cornea development in camera-type eye
hair cycle
catagen
anagen
positive regulation of apoptotic process
skin development
positive regulation of extrinsic apoptotic signaling pathway
molecular_function
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180536 | NM_001007461.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180536 | AY679090.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293663 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787017 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 861139 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 961744 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 221141 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206162 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 172598 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 148393 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159509 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133757 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180536 | NM_001007461.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180536 | AY679090.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28057521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28057519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28057518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28057517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28057524 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159509 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133757 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02406759 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159509 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133757 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09089152 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09089161 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09089158 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09300333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09116133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09116139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09134042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09456533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08994689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09456524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09456535 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10380959 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380961 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380964 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380960 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380963 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380962 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380957 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380958 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10380956 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100014598.mm.1 | NM_001007461.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014598.mm.1 | NM_001007461.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014598.mm.1 | TC1100001575.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014596.mm.1 | NM_001007461.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014596.mm.1 | NM_001007461.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014596.mm.1 | TC1100001575.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014597.mm.1 | NM_001007461.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014597.mm.1 | NM_001007461.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014597.mm.1 | TC1100001575.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161407 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161407 | - | - |
| mouse-6 v1.1 (Illumina) | GI_55925615-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3161407 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161407 | - | - |

Gsdma3
Genomic Map