Assay Details
| Gene Symbol: Zfp36l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009549.2 | NP_001009549.1 | 1 | 2240 | 78 | |
| GenBank mRNA | AY661338.1 | - | 1 | 2240 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp36l3
Genomic Map