Assay Details
| Gene Symbol: Nms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011684.2 | NP_001011684.3 | 6 - 7 | 481 | 168 | |
| GenBank mRNA | AB164466.1 | - | 6 - 7 | 481 | 168 | |
| BC138922.1 | - | 6 - 7 | 424 | 8861944 | 168 | |
| BQ175462.1 | - | 4 - 5 | 167 | |||
More Information
Set Membership: |
|

Nms
Genomic Map