Assay Details
| Gene Symbol: Nms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011684.2 | NP_001011684.3 | 9 - 10 | 608 | 147 | |
| GenBank mRNA | AB164466.1 | - | 9 - 10 | 608 | 147 | |
| BC138922.1 | - | 9 - 10 | 551 | 8861944 | 147 | |
| BQ175462.1 | - | 1 - 2 | 147 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Nms
Genomic Map