Assay Details
| Gene Symbol: Olfr963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011827.1 | NP_001011827.1 | 1 | 543 | 77 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2163473 | NM_001011827.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2163473 | NM_001011827.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439593 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439589 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439601 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26439628 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08193435 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08193447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26439630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08193437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07679177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07679170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08193460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07618370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08193431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26439630 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10584501 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10584500 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900002906.mm.1 | NM_001011827.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900002906.mm.1 | NM_001011827.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900002906.mm.1 | TC0900000433.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2959701 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2959701 | - | - |

Olfr963
Genomic Map