Assay Details
| Gene Symbol: Zbtb7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145356.3 | NP_663331.1 | 3 - 4 | 469 | 79 | |
| GenBank mRNA | AK045522.1 | - | 3 - 4 | 433 | 79 | |
| BC070424.1 | - | 3 - 4 | 469 | 6837447 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK045522.1, BC070424.1, NM_145356.3 transcripts.
Set Membership: |
|

Zbtb7c
Genomic Map