Assay Details
| Gene Symbol: Hecw2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172655.3 | NP_766243.2 | 7 | 1054 | 104 | |
| GenBank mRNA | AK042922.1 | - | 7 | 1055 | 104 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hecw2
Genomic Map