Assay Details
Gene Symbol: Gimap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031247.3 | NP_112537.2 | 5 | 1645 | 154 | |
GenBank mRNA | AB164418.1 | - | 5 | 1645 | 154 | |
AF337052.1 | - | 5 | 1652 | 156 | ||
AK008376.1 | - | 1 | 328 | 154 | ||
BC094914.1 | - | 5 | 1625 | 30074259 | 154 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003221 | BC094914.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003221 | NM_031247.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843410 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646820 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150001 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 39736 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579209 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84797 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175855 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003221 | BC094914.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003221 | NM_031247.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P516753 | AK008376.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P516753 | NM_031247.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P516753 | AB164418.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P516753 | AF337052.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429255 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429253 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429274 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429268 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429275 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429266 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429252 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429263 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429250 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429286 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429259 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429284 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429277 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429279 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429249 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429271 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429272 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429285 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429264 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429282 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24429256 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175855 | - | - |
Whole Mouse Genome G4122A | A_52_P374545 | AK008376.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P374545 | BC094914.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P374545 | NM_031247.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P374545 | AB164418.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P374545 | AF337052.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P516753 | AK008376.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P516753 | NM_031247.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P516753 | AB164418.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P516753 | AF337052.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113433 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175855 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05259786 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05259787 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05599598 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05254244 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05259784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24429256 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429243 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429240 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429234 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05175393 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05599599 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05420576 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429235 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429250 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429249 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429266 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05457745 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05175399 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05259787 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429283 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05259786 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429256 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429238 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429276 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429275 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429287 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429255 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05572219 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429245 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429220 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05098468 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05055864 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429222 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05599602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429271 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429259 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429216 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429268 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429237 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429279 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429224 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05457740 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429286 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429285 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429219 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429284 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429246 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429252 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429263 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24429253 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10544593 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544591 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544594 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544589 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544595 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544590 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544592 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10544588 | - | - |
Mouse Genome 430 2.0 Array | 1449220_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1449220_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 83408_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 83408_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600017523.mm.1 | TC0600002229.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600017523.mm.1 | NM_031247.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600017523.mm.1 | NM_031247.3 | Refseq |
Mouse Expression Set 430 | 1449220_PM_at | - | - |
Mouse Expression Set 430 | 1449220_at | - | - |
Murine Genome U74v2 Set | 170896_PM_at | - | - |
Murine Genome U74v2 Set | 170896_at | - | - |
Mouse Genome 430A 2.0 Array | 1449220_at | - | - |
Mouse Genome 430A 2.0 Array | 1449220_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 573645 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1740605 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1740605 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001740605 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1254241 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1254241 | - | - |
mouse-6 v1.1 (Illumina) | scl29043.6_66-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1740605 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl29043.6_66-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1740605 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1254241 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254241 | - | - |
