Assay Details
| Gene Symbol: Olfr884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011798.1 | NP_001011798.1 | 1 | 890 | 93 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2141898 | NM_001011798.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143179 | NM_001011798.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149351 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143179 | NM_001011798.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2141898 | NM_001011798.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433393 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433395 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433396 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149351 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149351 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07618302 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07618285 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07618313 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07618317 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07618312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26433415 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10584369 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584371 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10584368 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10584370 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161978 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161978 | - | - |
| mouse-6 v1.1 (Illumina) | GI_58801361-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3161978 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161978 | - | - |

Olfr884
Genomic Map