Assay Details
| Gene Symbol: Hdac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008228.2 | NP_032254.1 | 11 - 12 | 1232 | 67 | |
| GenBank mRNA | AK076080.1 | - | 11 - 12 | 1214 | 67 | |
| AK194630.1 | - | 1 - 2 | 41 | 67 | ||
| BC092070.1 | - | 11 - 12 | 1227 | 4217199 | 67 | |
| BC092285.1 | - | 11 - 12 | 1232 | 67 | ||
| BC108371.1 | - | 11 - 12 | 1233 | 4976514 | 67 | |
| U80780.1 | - | 11 - 12 | 1229 | 67 | ||
| X98207.1 | - | 11 - 12 | 1232 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hdac1
Genomic Map