Assay Details
| Gene Symbol: Ngfrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009750.2 | NP_033880.1 | 2 - 3 | 221 | 77 | |
| GenBank mRNA | AF097440.1 | - | 2 - 3 | 142 | 77 | |
| AF187066.1 | - | 2 - 3 | 147 | 77 | ||
| AK004531.1 | - | 2 - 3 | 151 | 77 | ||
| AK010500.1 | - | 2 - 3 | 148 | 77 | ||
| AK208231.1 | - | 2 - 3 | 155 | 77 | ||
| AV575702.1 | - | 2 - 3 | 222 | 77 | ||
| BC027815.1 | - | 2 - 3 | 147 | 5250429 | 77 | |
| CF618290.1 | - | 2 - 3 | 165 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF097440.1, AF187066.1, AK004531.1, AK010500.1, BC027815.1, NM_009750.2 transcripts.
Set Membership: |
|

Ngfrap1
Genomic Map