Assay Details
| Gene Symbol: Fhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010213.3 | NP_034343.2 | 2 | 225 | 77 | |
| GenBank mRNA | AF114382.1 | - | 2 | 196 | 77 | |
| AF134772.1 | - | 2 | 222 | 77 | ||
| AI426098.1 | - | 2 | 222 | 77 | ||
| BC144903.1 | - | 2 | 189 | 9053451 | 77 | |
| BC145939.1 | - | 2 | 189 | 40131242 | 77 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044448 | AF134772.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044448 | BC144903.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044448 | BC145939.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044448 | CB205184.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044448 | NM_010213.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028184 | AF149826.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028184 | BC144903.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028184 | BC145939.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028184 | CB205184.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028184 | NM_010213.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740264 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 667167 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 240171 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750301 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 21919 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123481 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044448 | AF134772.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044448 | BC144903.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044448 | BC145939.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044448 | CB205184.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044448 | NM_010213.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028184 | AF149826.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028184 | BC144903.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028184 | BC145939.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028184 | CB205184.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028184 | NM_010213.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P310635 | AF134772.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123481 | - | - |
| Whole Mouse Genome G4122A | A_51_P310635 | AF134772.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P65513 | AF149826.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P65513 | BC144903.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P65513 | BC145939.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P65513 | CB205184.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P65513 | NM_010213.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108360 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100465 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23269777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03600723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03215384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03325020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03325011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03757590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03745507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03884382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03884393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03884400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03215386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03325009 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03757591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03884402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03213167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03884387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03884399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03325019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03325015 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03884388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03884401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03884383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03325014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23269782 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10507912 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507910 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507911 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507913 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507909 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420779_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10507908 | - | - |
| Mouse Genome 430 2.0 Array | 1420779_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420779_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14201_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14201_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400010662.mm.1 | NM_010213.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010662.mm.1 | NM_010213.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010662.mm.1 | TC0400001419.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400010664.mm.1 | NM_010213.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010664.mm.1 | NM_010213.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010664.mm.1 | TC0400001419.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400010659.mm.1 | NM_010213.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010659.mm.1 | NM_010213.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010659.mm.1 | TC0400001419.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400010663.mm.1 | NM_010213.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010663.mm.1 | NM_010213.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010663.mm.1 | TC0400001419.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400010668.mm.1 | NM_010213.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010668.mm.1 | NM_010213.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010668.mm.1 | TC0400001419.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400010660.mm.1 | NM_010213.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010660.mm.1 | NM_010213.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010660.mm.1 | TC0400001419.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400010665.mm.1 | NM_010213.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010665.mm.1 | NM_010213.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010665.mm.1 | TC0400001419.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400010661.mm.1 | NM_010213.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010661.mm.1 | NM_010213.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400010661.mm.1 | TC0400001419.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420779_PM_at | - | - |
| Mouse Expression Set 430 | 1420779_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420779_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420779_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420779_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420779_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 860128 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2650017 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1090369 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2650017 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001090369 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002650017 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2708115 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2708115 | - | - |
| mouse-6 v1.1 (Illumina) | scl24989.2.50_2-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0002754.1_85-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2650017 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1090369 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002754.1_85-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl24989.2.50_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2650017 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1090369 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2708115 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2708115 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2686669 | - | - |

Fhl3
Genomic Map