Assay Details
| Gene Symbol: Vmn2r29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003555.1 | 7 | 904 | 61 | ||
| GenBank mRNA | AK018294.1 | - | 7 | 842 | 61 | |
| AK039785.1 | - | 1 | 2767 | 61 | ||
| BC096375.1 | - | 7 | 830 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374991 | BC096375.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374991 | AK039785.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374991 | NR_003555.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374991 | AK018294.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374991 | BC096375.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374991 | AK039785.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374991 | NR_003555.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P374991 | AK018294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P614860 | AK053659.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P614860 | AK084962.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374991 | BC096375.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374991 | AK039785.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374991 | NR_003555.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374991 | AK018294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374991 | BC096375.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374991 | AK039785.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374991 | NR_003555.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374991 | AK018294.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24963849 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10559913 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559843 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559845 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559893 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559842 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559841 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559848 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559935 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559849 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549915 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549920 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559934 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559838 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559932 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559891 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559911 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559840 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559844 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549918 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559839 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549922 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549931 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559915 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559881 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559892 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559930 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559847 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559850 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559846 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549914 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549919 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559933 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549943 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549916 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559910 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559912 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1445286_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453083_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559880 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10549938 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10549929 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10549921 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10549904 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559837 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559931 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10549917 | - | - |
| Mouse Genome 430 2.0 Array | 1453083_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1445286_at | - | - |
| Mouse Genome 430 2.0 Array | 1445286_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453083_at | - | - |
| Mouse Expression Set 430 | 1445286_at | - | - |
| Mouse Expression Set 430 | 1453083_at | - | - |
| Mouse Expression Set 430 | 1453083_PM_at | - | - |
| Mouse Expression Set 430 | 1445286_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453083_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445286_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453083_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445286_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 588227 | - | - |
| ABI Mouse Genome Survey Microarray | 830378 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 101660102 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106020537 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1252001 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213408 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1247517 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1252001 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106020537 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101660102 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101660102 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106020537 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213408 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1247517 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1252001 | - | - |

Vmn2r29
Genomic Map