Assay Details
Gene Symbol: Ifnz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197889.2 | NP_922871.1 | 1 | 1270 | 57 | |
GenBank mRNA | AK046550.1 | - | 1 | 1271 | 57 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
T cell activation involved in immune response
natural killer cell activation involved in immune response
humoral immune response
cytokine-mediated signaling pathway
B cell differentiation
positive regulation of peptidyl-serine phosphorylation of STAT protein
B cell proliferation
response to exogenous dsRNA
regulation of cell cycle
apoptotic signaling pathway
cytokine activity
type I interferon receptor binding
adaptive immune response
T cell activation involved in immune response
natural killer cell activation involved in immune response
humoral immune response
cytokine-mediated signaling pathway
B cell differentiation
positive regulation of peptidyl-serine phosphorylation of STAT protein
B cell proliferation
response to exogenous dsRNA
regulation of cell cycle
apoptotic signaling pathway
cytokine activity
type I interferon receptor binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Whole Mouse Genome G4122A | A_52_P471046 | AK144774.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P471046 | NM_001085532.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P471046 | NM_001098840.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P471046 | NM_001098841.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P471046 | NM_001085533.2 | mouse_transcriptome |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10505861 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505865 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505878 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10514330 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505857 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505875 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505849 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10514328 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505873 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10514332 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505859 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505854 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505867 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505844 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505851 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505860 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505853 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505855 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505871 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505869 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505877 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505847 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505863 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10514326 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505850 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10505846 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1427216_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10505872 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10505874 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10514327 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10505858 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10505848 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10505876 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10505868 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10514329 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10505852 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10505862 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10505856 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10505870 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10505845 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10514331 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10505864 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10505866 | - | - |
Mouse Genome 430 2.0 Array | 1427216_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1427216_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400006064.mm.1 | NM_197889.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400006064.mm.1 | NM_197889.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400006064.mm.1 | TC0400000895.mm.1 | MTA TC |
Mouse Expression Set 430 | 1427216_PM_at | - | - |
Mouse Expression Set 430 | 1425322_at | - | - |
Mouse Expression Set 430 | 1427216_at | - | - |
Mouse Expression Set 430 | 1425322_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1427216_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1425322_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1425322_at | - | - |
Mouse Genome 430A 2.0 Array | 1427216_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427216_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427216_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 106380048 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 106380048 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2480019 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002480019 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1247230 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2462559 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1247230 | - | - |
mouse-6 v1.1 (Illumina) | scl0319146.4_174-S | - | - |
mouse-6 v1.1 (Illumina) | scl00319146.2_327-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2480019 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 106380048 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00319146.2_327-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106380048 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2480019 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3093840 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3021483 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3093840 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3021483 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2462559 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1230408 | - | - |
