Assay Details
| Gene Symbol: H2-Ab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK155573.1 | - | 3 | 443 | 140 | |
| AK156899.1 | - | 3 | 443 | 140 | ||
| AK171005.1 | - | 3 | 443 | 140 | ||
| AK171211.1 | - | 3 | 447 | 140 | ||
| AK171281.1 | - | 3 | 443 | 140 | ||
| AK171290.1 | - | 3 | 443 | 140 | ||
| AK171298.1 | - | 3 | 443 | 140 | ||
| AK172409.1 | - | 3 | 444 | 140 | ||
| AY452202.1 | - | 3 | 380 | 140 | ||
| BC008168.1 | - | 3 | 463 | 3257494 | 140 | |
| BC010322.1 | - | 3 | 437 | 2651058 | 140 | |
| M15848.1 | - | 3 | 416 | 140 | ||
More Information
Set Membership: |
|

H2-Ab1
Genomic Map