Assay Details
| Gene Symbol: Rpl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016738.5 | NP_058018.2 | 6 | 710 | 87 | |
| GenBank mRNA | AK002787.1 | - | 5 | 647 | 87 | |
| AK198076.1 | - | 3 | 270 | 87 | ||
| AK202315.1 | - | 3 | 270 | 87 | ||
| AK204248.1 | - | 3 | 271 | 87 | ||
| BC034449.1 | - | 5 | 625 | 87 | ||
| BC055358.1 | - | 5 | 623 | 5352468 | 87 | |
| BC083148.1 | - | 3 | 1922 | 87 | ||
| CO039212.1 | - | 1 | 87 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26237921 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26237920 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26237918 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26237922 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991151 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009588 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011235 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 937408 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142528 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009588 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991151 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2032558 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011235 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P668076 | CO039212.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P668076 | U28917.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P668076 | BC083148.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P668076 | AK198076.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P668076 | AK202315.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P103513 | BC083148.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203420 | CO039212.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203420 | U28917.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203420 | BC083148.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203420 | BC085493.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203420 | CF751804.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26237909 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142528 | - | - |
| Whole Mouse Genome G4122A | A_52_P103513 | BC083148.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P668076 | CO039212.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P668076 | U28917.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P668076 | BC083148.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P668076 | AK198076.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P668076 | AK202315.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P203420 | CO039212.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P203420 | U28917.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P203420 | BC083148.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P203420 | BC085493.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P203420 | CF751804.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122498 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130471 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135683 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115910 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142528 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07374454 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07374434 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07374453 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07312242 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07312239 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26237918 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26237922 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26237921 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26237920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26237972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06802294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06802299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07245638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06802297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06706287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07374442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07018135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07312242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26237917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06843546 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10440662 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10472588 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531341 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576215 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576214 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576213 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379876 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501180 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415661 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576211 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576217 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10440663 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379877 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576212 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503250 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10492230 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455245_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10415660 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10503249 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10576210 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10576216 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10440661 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10379875 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10492229 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10501179 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10531340 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10472587 | - | - |
| Mouse Genome 430 2.0 Array | 1450150_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1455871_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1455871_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1455245_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1450150_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1460581_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1455245_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1460581_a_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800012310.mm.1 | NM_016738.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012310.mm.1 | NM_016738.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012310.mm.1 | TC0800001508.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800012306.mm.1 | NM_016738.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012306.mm.1 | NM_016738.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012306.mm.1 | TC0800001508.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800012298.mm.1 | NM_016738.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012298.mm.1 | NM_016738.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012298.mm.1 | TC0800001508.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800012297.mm.1 | NM_016738.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012297.mm.1 | NM_016738.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012297.mm.1 | TC0800001508.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800012293.mm.1 | NM_016738.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012293.mm.1 | NM_016738.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012293.mm.1 | TC0800001508.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800012304.mm.1 | NM_016738.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012304.mm.1 | NM_016738.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012304.mm.1 | TC0800001508.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800012311.mm.1 | NM_016738.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012311.mm.1 | NM_016738.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012311.mm.1 | TC0800001508.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455245_PM_x_at | - | - |
| Mouse Expression Set 430 | 1460581_a_at | - | - |
| Mouse Expression Set 430 | 1455871_s_at | - | - |
| Mouse Expression Set 430 | 1450150_a_at | - | - |
| Mouse Expression Set 430 | 1455871_PM_s_at | - | - |
| Mouse Expression Set 430 | 1450150_PM_a_at | - | - |
| Mouse Expression Set 430 | 1460581_PM_a_at | - | - |
| Mouse Expression Set 430 | 1455245_x_at | - | - |
| Murine Genome U74v2 Set | 102109_PM_at | - | - |
| Murine Genome U74v2 Set | 102109_at | - | - |
| Murine Genome U74v2 Set | 169415_PM_at | - | - |
| Murine Genome U74v2 Set | 169415_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455245_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450150_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460581_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455871_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455871_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450150_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460581_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455245_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 904555 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 580280 | AK030655.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 580280 | AK030655.1 | genbank_mrna_mouse |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000580280 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2734921 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2734921 | - | - |
| mouse-6 v1.1 (Illumina) | scl0270106.4_75-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 580280 | AK030655.1 | genbank_mrna_mouse |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0270106.4_75-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 580280 | AK030655.1 | genbank_mrna_mouse |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2734921 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0270106.4_75-S | - | - |

Rpl13
Genomic Map