Assay Details
| Gene Symbol: Lce1m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025420.2 | NP_079696.1 | 2 | 753 | 98 | |
| GenBank mRNA | AK003825.1 | - | 2 | 749 | 98 | |
| AK004318.1 | - | 2 | 755 | 98 | ||
| AK019517.1 | - | 2 | 753 | 98 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170059 | AK004318.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170059 | AK004318.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110255 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170059 | AK004318.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170059 | AK004318.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22334251 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110255 | - | - |
| Whole Mouse Genome G4122A | A_51_P170059 | AK004318.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170059 | AK004318.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134997 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110255 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02664356 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02664364 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02664352 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02664368 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02664351 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02664357 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02664372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22334321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02664358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334326 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02664372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334307 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02664351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334261 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02898279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334322 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03141503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02562314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02664366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02664356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02898288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334329 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334256 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334315 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334306 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334313 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02552300 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03141508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02664354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22334244 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10499957 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499956 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499955 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429565_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10499954 | - | - |
| Mouse Genome 430 2.0 Array | 1429565_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1429565_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66203_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66203_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300018672.mm.1 | NM_025420.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018672.mm.1 | NM_025420.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018672.mm.1 | TC0300002419.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429565_s_at | - | - |
| Mouse Expression Set 430 | 1429565_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429565_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429565_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 447270 | - | - |
| ABI Mouse Genome Survey Microarray | 743926 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5570435 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005570435 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2707612 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2707612 | - | - |
| mouse-6 v1.1 (Illumina) | scl21880.2_327-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5570435 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21880.2_327-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5570435 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2707612 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2707612 | - | - |

Lce1m
Genomic Map