Assay Details
| Gene Symbol: Acer2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK046888.1 | - | 2757 | 73 | ||
| BC051923.1 | - | 2717 | 73 | |||
| BC060063.1 | - | 2717 | 73 | |||
| BC061463.1 | - | 2713 | 73 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Acer2
Genomic Map