Assay Details
| Gene Symbol: 6330549D23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003619.2 | 2 | 2677 | 147 | ||
| GenBank mRNA | AK032024.1 | - | 2 | 2677 | 147 | |
| AK088233.1 | - | 3 | 3077 | 147 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ankyrin repeat domain 34A | Ankrd34a |
AW060394, Ankrd34 |
-6.5kb | Anti-sense | Chr. 3 - 96594826 - 96600126 on NCBI Build GRCm38 |
| Downstream | RNA binding motif protein 8a | Rbm8a |
2310057C03Rik, AA673428, Rbm8 |
22.6kb | Anti-sense | Chr. 3 - 96629928 - 96633791 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6330549D23Rik
Genomic Map