Assay Details
| Gene Symbol: Ifnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199157.2 | NP_954608.1 | 1 | 379 | 60 | |
| GenBank mRNA | BC099376.1 | - | 1 | 379 | 30713338 | 60 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
T cell activation involved in immune response
natural killer cell activation involved in immune response
regulation of transcription, DNA-templated
defense response
humoral immune response
cytokine-mediated signaling pathway
B cell differentiation
positive regulation of peptidyl-serine phosphorylation of STAT protein
B cell proliferation
response to exogenous dsRNA
defense response to virus
cytokine activity
cytokine receptor binding
type I interferon receptor binding
adaptive immune response
T cell activation involved in immune response
natural killer cell activation involved in immune response
regulation of transcription, DNA-templated
defense response
humoral immune response
cytokine-mediated signaling pathway
B cell differentiation
positive regulation of peptidyl-serine phosphorylation of STAT protein
B cell proliferation
response to exogenous dsRNA
defense response to virus
cytokine activity
cytokine receptor binding
type I interferon receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156588 | NM_199157.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156588 | BC099376.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167191 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106630 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162893 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156588 | NM_199157.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156588 | BC099376.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P128692 | NM_199157.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P128692 | BC099376.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22834521 | - | - |
| Whole Mouse Genome G4122A | A_52_P128692 | NM_199157.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P128692 | BC099376.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110754 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03772963 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04093147 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03772957 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03772964 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03772967 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03772961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22834582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834548 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834552 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834496 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834516 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834547 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03247155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834536 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03783793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03772965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834542 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834540 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03777013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04093145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04093143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03783796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22834546 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10503953 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503954 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10503952 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400002003.mm.1 | NM_199157.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400002003.mm.1 | NM_199157.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400002003.mm.1 | TC0400000317.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400002002.mm.1 | NM_199157.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400002002.mm.1 | NM_199157.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400002002.mm.1 | TC0400000317.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 316916 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5690064 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005690064 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2636143 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2636143 | - | - |
| mouse-6 v1.1 (Illumina) | scl0387510.1_98-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5690064 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0387510.1_98-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5690064 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2636143 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2636143 | - | - |

Ifnk
Genomic Map