Assay Details
| Gene Symbol: Sox12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011438.2 | NP_035568.1 | 1 | 4218 | 117 | |
| GenBank mRNA | AK081438.1 | - | 1 | 1340 | 117 | |
| BC067019.1 | - | 2 | 3955 | 6833848 | 117 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
spinal cord development
positive regulation of transcription from RNA polymerase II promoter
protein-DNA complex assembly
transcription regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription coactivator activity
DNA binding
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
spinal cord development
positive regulation of transcription from RNA polymerase II promoter
protein-DNA complex assembly
transcription regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription coactivator activity
DNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749129 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749123 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749119 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749121 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749128 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749117 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749126 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749133 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749130 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749118 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749127 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749134 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749124 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749131 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749116 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749122 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749132 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P115541 | AK081438.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P115541 | BC067019.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P115541 | NM_011438.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 45975 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 20273 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171620 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P115541 | AK081438.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P115541 | BC067019.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P115541 | NM_011438.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P322450 | Z18961.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749126 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171620 | - | - |
| Whole Mouse Genome G4122A | A_51_P322450 | Z18961.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171620 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02224545 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02224546 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02224542 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02224536 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02224539 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749117 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749118 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749128 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749130 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749127 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749119 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749121 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749126 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749129 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749122 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749132 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749133 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749116 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749124 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749123 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749131 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21749134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01744379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01424415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01205804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02077384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01424405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01972831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01424422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02229005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02224547 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01424428 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01424419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01699841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02224544 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01652864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02224536 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01205808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02077382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01385020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02224542 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01385017 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01424407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01424416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02224539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02224545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01652860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01385012 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10488624 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488625 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439754_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455562_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456882_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10488623 | - | - |
| Mouse Genome 430 2.0 Array | 1439754_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455562_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456882_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456882_at | - | - |
| Mouse Genome 430 2.0 Array | 1455562_at | - | - |
| Mouse Genome 430 2.0 Array | 1439754_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20667_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20667_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200041607.mm.1 | NM_011438.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041607.mm.1 | NM_011438.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041607.mm.1 | TC0200004898.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200041608.mm.1 | NM_011438.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041608.mm.1 | NM_011438.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041608.mm.1 | TC0200004898.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455562_PM_at | - | - |
| Mouse Expression Set 430 | 1455562_at | - | - |
| Mouse Expression Set 430 | 1456882_at | - | - |
| Mouse Expression Set 430 | 1456882_PM_at | - | - |
| Mouse Expression Set 430 | 1439754_PM_at | - | - |
| Mouse Expression Set 430 | 1439754_at | - | - |
| Murine Genome U74v2 Set | 138465_at | - | - |
| Murine Genome U74v2 Set | 138465_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455562_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455562_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455562_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455562_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456882_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439754_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439754_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456882_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 527938 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1780037 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102900040 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001780037 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2775281 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1227128 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2775281 | - | - |
| mouse-6 v1.1 (Illumina) | scl069985.1_46-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0020667.1_65-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102900040 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1780037 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0020667.1_65-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102900040 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1780037 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3026635 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3099470 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3026635 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1227128 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3099470 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2775281 | - | - |

Sox12
Genomic Map