Assay Details
| Gene Symbol: Ube3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033962.1 | NP_001029134.1 | 3 - 4 | 851 | 134 | |
| NM_011668.2 | NP_035798.2 | 5 - 6 | 1038 | 134 | ||
| NM_173010.3 | NP_766598.1 | 3 - 4 | 853 | 134 | ||
| GenBank mRNA | AK029133.1 | - | 3 - 4 | 854 | 134 | |
| AK083795.1 | - | 3 - 4 | 782 | 134 | ||
| AK220549.1 | - | 3 - 4 | 751 | 134 | ||
| BC131658.1 | - | 3 - 4 | 839 | 40110636 | 134 | |
| BC131659.1 | - | 3 - 4 | 839 | 40110637 | 134 | |
| U82122.1 | - | 1 - 2 | 288 | 134 | ||
| U96636.1 | - | 5 - 6 | 585 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ube3a
Genomic Map