Assay Details
| Gene Symbol: H2-T3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY989874.1 | - | 3 - 4 | 620 | 90 | |
| AY989875.1 | - | 3 - 4 | 603 | 90 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2-T3| Gene Symbol: H2-T3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY989874.1 | - | 3 - 4 | 620 | 90 | |
| AY989875.1 | - | 3 - 4 | 603 | 90 | ||
Set Membership: |
|