Assay Details
| Gene Symbol: Smarcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025891.3 | NP_080167.3 | 5 - 6 | 728 | 120 | |
| GenBank mRNA | AK005094.1 | - | 5 - 6 | 675 | 120 | |
| BC013122.1 | - | 5 - 6 | 765 | 3586276 | 120 | |
| BC060525.1 | - | 5 - 6 | 627 | 6817219 | 120 | |
More Information
Set Membership: |
|

Smarcd3
Genomic Map