Assay Details
| Gene Symbol: Vwde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013757.2 | NP_001013779.2 | 9 - 10 | 1432 | 93 | |
| GenBank mRNA | BC076621.1 | - | 9 - 10 | 1432 | 93 | |
| BC151200.1 | - | 9 - 10 | 1341 | 9088088 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vwde
Genomic Map