Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800020616.mm.1 |
NM_001013764.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020616.mm.1 |
NM_001013764.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020616.mm.1 |
TC0800002698.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020614.mm.1 |
NM_001013764.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020614.mm.1 |
NM_001013764.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020614.mm.1 |
TC0800002698.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020615.mm.1 |
NM_001013764.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020615.mm.1 |
NM_001013764.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020615.mm.1 |
TC0800002698.mm.1 |
MTA TC |