Assay Details
| Gene Symbol: Igsf9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033323.3 | NP_001028495.2 | 11 - 12 | 1746 | 89 | |
| NM_001129787.1 | NP_001123259.1 | 11 - 12 | 1746 | 89 | ||
| GenBank mRNA | AK158950.1 | - | 11 - 12 | 1746 | 89 | |
| BC150882.1 | - | 11 - 12 | 1554 | 89 | ||
| BC150887.1 | - | 11 - 12 | 1554 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igsf9b
Genomic Map