Assay Details
| Gene Symbol: Krt26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033397.5 | NP_001028569.2 | 4 - 5 | 891 | 77 | |
| GenBank mRNA | AK028591.1 | - | 4 - 5 | 891 | 77 | |
| AK028682.1 | - | 3 - 4 | 2024 | 77 | ||
| AK132445.1 | - | 4 - 5 | 895 | 77 | ||
| AK162718.1 | - | 4 - 5 | 893 | 77 | ||
| BC116672.1 | - | 4 - 5 | 880 | 40107596 | 77 | |
More Information
Set Membership: |
|

Krt26
Genomic Map