Assay Details
| Gene Symbol: Bsph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033418.4 | NP_001028590.1 | 3 - 4 | 202 | 154 | |
| GenBank mRNA | AK136662.1 | - | 3 - 4 | 203 | 154 | |
| BC147454.1 | - | 3 - 4 | 187 | 9056096 | 154 | |
| BC147459.1 | - | 3 - 4 | 187 | 9056101 | 154 | |
| DQ227498.1 | - | 3 - 4 | 182 | 154 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bsph1
Genomic Map