Assay Details
| Gene Symbol: Gm1527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033479.4 | NP_001028651.1 | 9 - 10 | 1068 | 64 | |
| GenBank mRNA | AK132999.1 | - | 9 - 10 | 1067 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2030692 | NM_001033479.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2030692 | AK132999.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481466 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 24098 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2030692 | NM_001033479.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2030692 | AK132999.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00611193 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04863753 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00611189 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00611183 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04863702 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04863753 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00611193 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00611177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22034131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22034113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22034125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22034256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22034250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22034134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22034247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22034143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32981626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32981581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32981610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32981586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32981629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32981594 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32981628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32981572 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P32981593 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10491199 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491203 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491207 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491205 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491202 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491206 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491209 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491198 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491201 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491211 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491204 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491208 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491200 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491210 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10491197 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300000993.mm.1 | NM_001033479.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000993.mm.1 | NM_001033479.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000993.mm.1 | TC0300000185.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300000992.mm.1 | NM_001033479.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000992.mm.1 | NM_001033479.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000992.mm.1 | TC0300000185.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300000990.mm.1 | NM_001033479.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000990.mm.1 | NM_001033479.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000990.mm.1 | TC0300000185.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300000991.mm.1 | NM_001033479.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000991.mm.1 | NM_001033479.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000991.mm.1 | TC0300000185.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300000994.mm.1 | NM_001033479.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000994.mm.1 | NM_001033479.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000994.mm.1 | TC0300000185.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300000989.mm.1 | NM_001033479.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000989.mm.1 | NM_001033479.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000989.mm.1 | TC0300000185.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1248201 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3064066 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3064066 | - | - |

Gm1527
Genomic Map