Assay Details
| Gene Symbol: Slc47a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033542.2 | NP_001028714.1 | 15 - 16 | 1516 | 76 | |
| GenBank mRNA | AK133432.1 | - | 15 - 16 | 1517 | 76 | |
| BC147619.1 | - | 15 - 16 | 1464 | 9056609 | 76 | |
More Information
Set Membership: |
|

Slc47a2
Genomic Map