Assay Details
| Gene Symbol: Ccdc185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033547.2 | NP_001028719.1 | 1 | 1788 | 87 | |
| GenBank mRNA | AK077097.1 | - | 1 | 1096 | 87 | |
| AK132754.1 | - | 1 | 1789 | 87 | ||
| BC150917.1 | - | 1 | 1759 | 9087828 | 87 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P303696 | AK132754.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P303696 | BC150917.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P303696 | NM_001033547.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P303696 | AK077097.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P303696 | AK132754.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P303696 | BC150917.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P303696 | NM_001033547.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P303696 | AK077097.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 946999 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133713 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P303696 | AK132754.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P303696 | BC150917.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P303696 | NM_001033547.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P303696 | AK077097.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P303696 | AK132754.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P303696 | BC150917.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P303696 | NM_001033547.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P303696 | AK077097.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888641 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20888665 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133713 | - | - |
| Whole Mouse Genome G4122A | A_51_P303696 | AK132754.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P303696 | BC150917.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P303696 | NM_001033547.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P303696 | AK077097.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P303696 | AK132754.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P303696 | BC150917.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P303696 | NM_001033547.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P303696 | AK077097.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104106 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20888601 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00851931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00837563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888625 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00269221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00808761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00851922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00851921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00450708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00513404 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01132397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888601 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00461869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00419485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888612 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00851932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00850553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00048545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00851937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20888616 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10360829 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360830 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360831 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439230_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10360828 | - | - |
| Mouse Genome 430 2.0 Array | 1439230_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439230_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 433386_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 433386_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100030471.mm.1 | NM_001033547.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100030471.mm.1 | NM_001033547.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100030471.mm.1 | TC0100003719.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100030472.mm.1 | NM_001033547.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100030472.mm.1 | NM_001033547.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100030472.mm.1 | TC0100003719.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100030473.mm.1 | NM_001033547.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100030473.mm.1 | NM_001033547.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100030473.mm.1 | TC0100003719.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439230_at | - | - |
| Mouse Expression Set 430 | 1439230_PM_at | - | - |
| Murine Genome U74v2 Set | 166385_i_at | - | - |
| Murine Genome U74v2 Set | 166385_PM_i_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439230_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439230_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103140458 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103140458 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230466 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103140458 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103140458 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2825496 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2825496 | - | - |

Ccdc185
Genomic Map