Assay Details
| Gene Symbol: Dnaaf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033548.2 | NP_001028720.1 | 4 - 5 | 393 | 63 | |
| GenBank mRNA | AK134335.1 | - | 4 - 5 | 394 | 63 | |
| BC150983.1 | - | 4 - 5 | 379 | 9087896 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnaaf3
Genomic Map