Assay Details
| Gene Symbol: Dnaaf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033548.2 | NP_001028720.1 | 6 - 7 | 733 | 64 | |
| GenBank mRNA | AK134335.1 | - | 6 - 7 | 734 | 64 | |
| BC150983.1 | - | 6 - 7 | 719 | 9087896 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnaaf3
Genomic Map