Assay Details
| Gene Symbol: Stmn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019641.4 | NP_062615.1 | 3 - 4 | 353 | 145 | |
| GenBank mRNA | AB064953.1 | - | 3 - 4 | 306 | 145 | |
| AK002606.1 | - | 3 - 4 | 275 | 145 | ||
| AK012231.1 | - | 3 - 4 | 275 | 145 | ||
| AK012347.1 | - | 3 - 4 | 275 | 145 | ||
| AK013302.1 | - | 3 - 4 | 275 | 145 | ||
| AK014108.1 | - | 3 - 4 | 276 | 145 | ||
| BC010581.1 | - | 3 - 4 | 262 | 3154091 | 145 | |
| BC031831.1 | - | 3 - 4 | 261 | 4500926 | 145 | |
| BC054396.1 | - | 2 - 3 | 254 | 6492045 | 145 | |
| X94915.1 | - | 1 - 2 | 183 | 145 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Stmn1
Genomic Map