Assay Details
| Gene Symbol: Ppih | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110129.1 | NP_001103599.1 | 6 - 7 | 380 | 126 | |
| NM_001110130.1 | NP_001103600.1 | 6 - 7 | 380 | 126 | ||
| NM_028677.4 | NP_082953.1 | 6 - 7 | 380 | 126 | ||
| GenBank mRNA | AK003179.1 | - | 6 - 7 | 358 | 126 | |
| AK005202.1 | - | 6 - 7 | 341 | 126 | ||
| AK008394.1 | - | 6 - 7 | 358 | 126 | ||
| AK014665.1 | - | 6 - 7 | 341 | 126 | ||
| AK134995.1 | - | 6 - 7 | 360 | 126 | ||
| AK136330.1 | - | 6 - 7 | 339 | 126 | ||
| AK164299.1 | - | 6 - 7 | 345 | 126 | ||
| BC016565.1 | - | 6 - 7 | 335 | 126 | ||
| BC050116.1 | - | 6 - 7 | 334 | 6432881 | 126 | |
| BC098490.1 | - | 336 | 30022355 | 126 | ||
| BY708305.1 | - | 6 - 7 | 357 | 126 | ||
| Gene Symbol: Gm10069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028592.1 | 3 | 126 | |||
| GenBank mRNA | AK029950.1 | - | 3 | 126 | ||
| AK046088.1 | - | 3 | 126 | |||
| AK086937.1 | - | 3 | 126 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ppih
Genomic Map