Assay Details
| Gene Symbol: Serpina3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009251.1 | NP_033277.1 | 1119 | 116 | ||
| GenBank mRNA | AA210495.1 | - | 2 - 3 | 184 | 116 | |
| AK156462.1 | - | 4 - 5 | 1117 | 116 | ||
| BC002065.1 | - | 4 - 5 | 908 | 116 | ||
| BC057144.1 | - | 3 - 4 | 760 | 116 | ||
| BC096664.1 | - | 3 - 4 | 421 | 116 | ||
| BC119537.2 | - | 1135 | 40056419 | 116 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
immune system process
apoptotic process
negative regulation of peptidase activity
negative regulation of endopeptidase activity
response to cytokine
response to peptide hormone
serine-type endopeptidase inhibitor activity
cysteine-type endopeptidase inhibitor activity
peptidase inhibitor activity
immune system process
apoptotic process
negative regulation of peptidase activity
negative regulation of endopeptidase activity
response to cytokine
response to peptide hormone
serine-type endopeptidase inhibitor activity
cysteine-type endopeptidase inhibitor activity
peptidase inhibitor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P326191 | M64085.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P326191 | M64085.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 212480 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675608 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 123709 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644623 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 83397 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 326389 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 878273 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616455 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 557257 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211414 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 100994 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 14529 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158989 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155960 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P326191 | M64085.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P326191 | M64085.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P326191 | M64085.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P326191 | M64085.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702042 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158989 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155960 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07734097 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07734109 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02595005 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02595022 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07734089 | - | - |
| Whole Mouse Genome G4122A | A_51_P326191 | M64085.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P326191 | M64085.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07734109 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155960 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158989 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10076275 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10076284 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10076281 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10076283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09904063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10375489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10076284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09761863 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09761871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10036555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09816994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10375504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702153 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10276971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10375471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09904062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09761874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10419478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10375468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10276970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10354610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10070309 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10398047 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398049 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398050 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398046 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398048 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398051 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424923_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10398039 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10398052 | - | - |
| Mouse Genome 430 2.0 Array | 1424923_at | - | - |
| Mouse Genome 430 2.0 Array | 1424923_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20715_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20715_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa11780_at | - | - |
| Mouse Expression Set 430 | 1424923_PM_at | - | - |
| Mouse Expression Set 430 | 1424923_at | - | - |
| Murine Genome U74v2 Set | 102860_PM_at | - | - |
| Murine Genome U74v2 Set | 102860_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424923_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424923_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424923_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424923_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 485872 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 610600 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2725927 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2725927 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2725927 | - | - |

Serpina3g
Genomic Map