Assay Details
| Gene Symbol: Rbm12b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198957.2 | NP_945195.1 | 4 | 3230 | 138 | |
| GenBank mRNA | AK021260.1 | - | 1 | 263 | 138 | |
| AK080943.1 | - | 4 | 3158 | 138 | ||
| AK169895.1 | - | 2 | 2652 | 138 | ||
| BC030408.1 | - | 1 | 1567 | 138 | ||
| BC080741.1 | - | 3 | 30461213 | 138 | ||
| BY620336.1 | - | 1 | 188 | 138 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084128 | BC137620.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084128 | BC030408.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084128 | BC080741.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084128 | AK080943.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084128 | AK169895.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016739 | BB653094.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016739 | BC030408.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016739 | BC080741.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016739 | AK021260.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016739 | AK080943.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 647703 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134858 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016739 | BB653094.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016739 | BC030408.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016739 | BC080741.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016739 | AK021260.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016739 | AK080943.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084128 | BC137620.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084128 | BC030408.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084128 | BC080741.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084128 | AK080943.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084128 | AK169895.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22733198 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134858 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22733139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22733198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22733246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22733180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03284042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22733203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22733244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22733235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22733234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22733238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22733178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22733239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03244200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22733273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22733245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22733236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22733240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22733281 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10503369 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503368 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503365 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503364 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503360 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503366 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503367 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436843_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1442663_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10503363 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10503359 | - | - |
| Mouse Genome 430 2.0 Array | 1436843_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1442663_at | - | - |
| Mouse Genome 430 2.0 Array | 1442663_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436843_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 77604_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 77604_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400000712.mm.1 | NM_198957.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000712.mm.1 | NM_198957.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000712.mm.1 | TC0400000103.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400000711.mm.1 | NM_198957.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000711.mm.1 | NM_198957.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000711.mm.1 | TC0400000103.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400000716.mm.1 | NM_198957.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000716.mm.1 | NM_198957.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000716.mm.1 | TC0400000103.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400000719.mm.1 | NM_198957.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000719.mm.1 | NM_198957.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000719.mm.1 | TC0400000103.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400000709.mm.1 | NM_198957.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000709.mm.1 | NM_198957.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000709.mm.1 | TC0400000103.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400000717.mm.1 | NM_198957.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000717.mm.1 | NM_198957.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000717.mm.1 | TC0400000103.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400000715.mm.1 | NM_198957.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000715.mm.1 | NM_198957.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000715.mm.1 | TC0400000103.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400000721.mm.1 | NM_198957.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000721.mm.1 | NM_198957.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000721.mm.1 | TC0400000103.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400000718.mm.1 | NM_198957.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000718.mm.1 | NM_198957.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000718.mm.1 | TC0400000103.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400000720.mm.1 | NM_198957.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000720.mm.1 | NM_198957.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000720.mm.1 | TC0400000103.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400000714.mm.1 | NM_198957.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000714.mm.1 | NM_198957.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400000714.mm.1 | TC0400000103.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1442663_at | - | - |
| Mouse Expression Set 430 | 1442663_PM_at | - | - |
| Murine Genome U74v2 Set | 165652_r_at | - | - |
| Murine Genome U74v2 Set | 165652_PM_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436843_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436843_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442663_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442663_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 335161 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2679390 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2679390 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2679390 | - | - |

Rbm12b2
Genomic Map