Assay Details
| Gene Symbol: Adrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007420.3 | NP_031446.2 | 1 | 1881 | 145 | |
| GenBank mRNA | AK080276.1 | - | 1 | 146 | ||
| BC030346.1 | - | 1 | 1333 | 145 | ||
| BC032883.1 | - | 1 | 1859 | 1349283 | 145 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Adrb2
Genomic Map