Assay Details
| Gene Symbol: Fzd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008057.3 | NP_032083.3 | 1 | 4501 | 113 | |
| GenBank mRNA | AK010239.2 | - | 1 | 2082 | 113 | |
| BC049781.1 | - | 1 | 4221 | 113 | ||
| BC063077.1 | - | 2 | 3832 | 6844727 | 113 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fzd7
Genomic Map