| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400030253.mm.1 |
TC0400003691.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400030253.mm.1 |
NM_008154.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030253.mm.1 |
NM_008154.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030254.mm.1 |
TC0400003691.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400030254.mm.1 |
NM_008154.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030254.mm.1 |
NM_008154.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030252.mm.1 |
TC0400003691.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400030252.mm.1 |
NM_008154.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030252.mm.1 |
NM_008154.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030258.mm.1 |
TC0400003691.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400030258.mm.1 |
NM_008154.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030258.mm.1 |
NM_008154.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030255.mm.1 |
TC0400003691.mm.1 |
MTA TC |