Assay Details
| Gene Symbol: Gpr132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019925.4 | NP_064309.1 | 3 | 1746 | 136 | |
| GenBank mRNA | AK089866.1 | - | 3 | 1723 | 136 | |
| AK153765.1 | - | 3 | 1747 | 136 | ||
| AK157600.1 | - | 3 | 1773 | 136 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28753097 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28753098 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28753095 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28753094 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28753096 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051229 | AK153765.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051229 | AK154509.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051229 | NM_019925.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 294949 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 88361 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604888 | BC119215.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604888 | BC119217.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604888 | NM_207235.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204409 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727696 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425518 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148533 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 243533 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 151161 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687781 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906575 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 192147 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 135080 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 120135 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 3018 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 87690 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29327 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133594 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051229 | AK153765.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051229 | AK154509.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051229 | NM_019925.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177340 | AK157600.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177340 | BC120522.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177340 | BC120524.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177340 | AK153765.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177340 | AK154509.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07755345 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133594 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07755336 | - | - |
| Whole Mouse Genome G4122A | A_51_P177340 | AK157600.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177340 | BC120522.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177340 | BC120524.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177340 | AK153765.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177340 | AK154509.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140735 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133594 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10206149 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10206147 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10206135 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10206143 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10206146 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10166309 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28753094 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28753097 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28753098 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28753096 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28753095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28752910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28753017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28753023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10291011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28753004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28753007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10420249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28753002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28753012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10227896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752965 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10291012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09853859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28753017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28753011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28753014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09873060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28753009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10420246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28753025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10420224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28753006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28753010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10166314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10213940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10370352 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10291010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28752977 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10402806 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402805 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402807 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402802 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402801 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402804 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402803 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444233_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421755_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10402800 | - | - |
| Mouse Genome 430 2.0 Array | 1444233_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421755_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421755_at | - | - |
| Mouse Genome 430 2.0 Array | 1444233_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56696_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56696_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200016953.mm.1 | NM_019925.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016953.mm.1 | NM_019925.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016953.mm.1 | TC1200002440.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200016956.mm.1 | NM_019925.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016956.mm.1 | NM_019925.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016956.mm.1 | TC1200002440.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200016955.mm.1 | NM_019925.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016955.mm.1 | NM_019925.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016955.mm.1 | TC1200002440.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200016951.mm.1 | NM_019925.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016951.mm.1 | NM_019925.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016951.mm.1 | TC1200002440.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200016954.mm.1 | NM_019925.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016954.mm.1 | NM_019925.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016954.mm.1 | TC1200002440.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421755_at | - | - |
| Mouse Expression Set 430 | 1421755_PM_at | - | - |
| Mouse Expression Set 430 | 1444233_PM_at | - | - |
| Mouse Expression Set 430 | 1444233_at | - | - |
| Murine Genome U74v2 Set | 94566_at | - | - |
| Murine Genome U74v2 Set | 94566_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421755_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421755_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421755_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421755_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444233_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444233_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 435670 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4150114 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004150114 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2659578 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2659578 | - | - |
| mouse-6 v1.1 (Illumina) | scl056696.1_260-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4150114 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl056696.1_260-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4150114 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2659578 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2659578 | - | - |

Gpr132
Genomic Map