Assay Details
| Gene Symbol: Gpr18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182806.1 | NP_877958.1 | 2 | 1145 | 139 | |
| GenBank mRNA | AK156039.1 | - | 2 | 1218 | 139 | |
| AK156868.1 | - | 2 | 1210 | 139 | ||
| AK157029.1 | - | 2 | 1153 | 139 | ||
| AK172401.1 | - | 2 | 1148 | 139 | ||
| BC034872.1 | - | 2 | 1145 | 1397739 | 139 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P216613 | BC034872.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P216613 | AK172401.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P216613 | NM_182806.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P216613 | AK156039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P216613 | AK156868.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P216613 | BC034872.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P216613 | AK172401.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P216613 | NM_182806.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P216613 | AK156039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P216613 | AK156868.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104680 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P216613 | BC034872.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P216613 | AK172401.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P216613 | NM_182806.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P216613 | AK156039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P216613 | AK156868.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P216613 | BC034872.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P216613 | AK172401.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P216613 | NM_182806.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P216613 | AK156039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P216613 | AK156868.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P216613 | BC034872.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P216613 | AK172401.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P216613 | NM_182806.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P216613 | AK156039.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P216613 | AK156868.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P216613 | BC034872.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P216613 | AK172401.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P216613 | NM_182806.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P216613 | AK156039.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P216613 | AK156868.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958601 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958589 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29958561 | - | - |
| Whole Mouse Genome G4122A | A_52_P216613 | BC034872.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P216613 | AK172401.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P216613 | NM_182806.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P216613 | AK156039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P216613 | AK156868.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P216613 | BC034872.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P216613 | AK172401.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P216613 | NM_182806.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P216613 | AK156039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P216613 | AK156868.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123579 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11615575 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11436518 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11615579 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11615577 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11436519 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11436516 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11615576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29958578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958611 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11141454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11410469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11141445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11164108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11164105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11164106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958586 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958609 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958563 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958621 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11282380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958542 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958561 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958627 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11299127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11141444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29958579 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10422494 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10422495 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439141_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10422493 | - | - |
| Mouse Genome 430 2.0 Array | 1439141_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439141_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 110168_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 110168_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400020472.mm.1 | NM_182806.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400020472.mm.1 | NM_182806.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400020472.mm.1 | TC1400002744.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400020473.mm.1 | NM_182806.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400020473.mm.1 | NM_182806.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400020473.mm.1 | TC1400002744.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439141_PM_at | - | - |
| Mouse Expression Set 430 | 1439141_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439141_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439141_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 522671 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2640551 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002640551 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213809 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213809 | - | - |
| mouse-6 v1.1 (Illumina) | scl45134.2_536-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2640551 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl45134.2_536-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2640551 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1213809 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213809 | - | - |

Gpr18
Genomic Map