Assay Details
| Gene Symbol: Gpr152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206973.2 | NP_996856.1 | 2 | 3540 | 117 | |
| GenBank mRNA | BC099681.1 | - | 2 | 3540 | 6494327 | 117 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051236 | BC099681.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2051236 | NM_206973.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 52463 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 839501 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 548929 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329210 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96464 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109822 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051236 | BC099681.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2051236 | NM_206973.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P1093503 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519524 | BC099681.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519524 | AK044366.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519524 | NM_206973.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921329 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109822 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03540675 | - | - |
| Whole Mouse Genome G4122A | A_51_P519524 | BC099681.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519524 | AK044366.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519524 | NM_206973.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1093503 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102752 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109822 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14253605 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14253614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14253613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14426358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921339 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14474848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14426370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921328 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14253614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921341 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14426349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14426360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921344 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921346 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921316 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921324 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10460357 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460358 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438574_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10460356 | - | - |
| Mouse Genome 430 2.0 Array | 1438574_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438574_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269053_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269053_PM_at | - | - |
| Mouse Expression Set 430 | 1438574_PM_at | - | - |
| Mouse Expression Set 430 | 1438574_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438574_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438574_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 616524 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6200025 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6200025 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006200025 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240344 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240344 | - | - |
| mouse-6 v1.1 (Illumina) | scl0269053.1_197-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6200025 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0269053.1_197-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6200025 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1240344 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240344 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0269053.1_197-S | - | - |

Gpr152
Genomic Map