Assay Details
Gene Symbol: Gm6878 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037931.3 | NP_001033020.1 | 3 - 4 | 613 | 92 | |
GenBank mRNA | AK134891.1 | - | 3 - 4 | 615 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2185955 | NM_001037931.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2185955 | AK134891.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2185955 | NM_001037931.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2185955 | AK134891.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684209 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29684195 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684296 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684314 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684258 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684306 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684269 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684278 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684275 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684344 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684223 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684293 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684245 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684217 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11198010 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684263 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684287 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11784196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684198 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684207 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684206 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684302 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684273 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11312910 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684331 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11681022 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684192 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684227 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684266 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684308 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684234 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684199 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684274 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684219 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684230 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684209 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11822423 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684255 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684300 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684311 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684214 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684228 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684254 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684246 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684210 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11198015 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11601815 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684243 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684272 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11832469 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684225 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684326 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684268 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684253 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684250 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684211 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684216 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684205 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684221 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684339 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684195 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684341 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684320 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684318 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11736780 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11631033 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684332 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684289 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684236 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684193 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11822414 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684232 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684265 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684313 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684251 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684244 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684262 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684337 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684284 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684213 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684237 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684333 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684239 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684338 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11299677 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684218 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684335 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684220 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684212 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684343 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29684298 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10421025 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10421024 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10421022 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10421023 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10421021 | - | - |
Affymetrix Mouse Clariom S Array | PSR1400017649.mm.1 | NM_001037931.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400017649.mm.1 | NM_001037931.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400017649.mm.1 | TC1400002286.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1224350 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2839710 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2839710 | - | - |
