Assay Details
| Gene Symbol: Defb26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039120.2 | NP_001034209.1 | 1 - 2 | 104 | 95 | |
| GenBank mRNA | BC139131.1 | - | 1 - 2 | 104 | 8862153 | 95 |
| DQ012037.1 | - | 1 - 2 | 60 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974377 | NM_001039120.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974377 | BC139131.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974377 | DQ012037.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795997 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 709326 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 104962 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103584 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974377 | NM_001039120.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974377 | BC139131.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974377 | DQ012037.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103584 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01491088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749637 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01491089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01955538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01519377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01432560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01491079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01344540 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01172949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01761662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01770332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01193716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21749652 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10488638 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488637 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10488636 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200041627.mm.1 | NM_001039120.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041627.mm.1 | NM_001039120.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041627.mm.1 | TC0200004902.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200041625.mm.1 | NM_001039120.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041625.mm.1 | NM_001039120.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041625.mm.1 | TC0200004902.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200041624.mm.1 | NM_001039120.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041624.mm.1 | NM_001039120.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041624.mm.1 | TC0200004902.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200041626.mm.1 | NM_001039120.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041626.mm.1 | NM_001039120.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041626.mm.1 | TC0200004902.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161354 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161354 | - | - |
| mouse-6 v1.1 (Illumina) | GI_84993769-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3161354 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161354 | - | - |

Defb26
Genomic Map