Assay Details
| Gene Symbol: Spag11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039563.3 | NP_001034652.2 | 2 - 3 | 251 | 118 | |
| NM_001286493.1 | NP_001273422.1 | 2 - 3 | 254 | 118 | ||
| GenBank mRNA | AY552530.1 | - | 2 - 3 | 217 | 118 | |
| BC145547.1 | - | 2 - 3 | 251 | 9054111 | 118 | |
| BC145549.1 | - | 2 - 3 | 248 | 9054113 | 118 | |
| BC145818.1 | - | 2 - 3 | 248 | 40131121 | 118 | |
| BC145820.1 | - | 2 - 3 | 248 | 40131123 | 118 | |
| DQ012055.1 | - | 2 - 3 | 212 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171768 | AK136627.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171768 | NM_001034905.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171764 | AK136627.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171764 | AY552530.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171764 | NM_001034905.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171764 | NM_001039563.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515271 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 140314 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 123281 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171768 | AK136627.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171768 | NM_001034905.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171764 | AK136627.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171764 | AY552530.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171764 | NM_001034905.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171764 | NM_001039563.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702145 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06443201 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06443201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06882953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06882955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06882923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06882925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06882913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06882952 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10570595 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570596 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570593 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570597 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570594 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570598 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570591 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570590 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800001571.mm.1 | NM_001034905.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001571.mm.1 | NM_001034905.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001571.mm.1 | NM_001039563.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001571.mm.1 | NM_001286493.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001571.mm.1 | TC0800000177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800001570.mm.1 | NM_001034905.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001570.mm.1 | NM_001034905.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001570.mm.1 | NM_001039563.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001570.mm.1 | NM_001286493.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001570.mm.1 | TC0800000177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800001569.mm.1 | NM_001034905.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001569.mm.1 | NM_001034905.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001569.mm.1 | NM_001039563.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001569.mm.1 | NM_001286493.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001569.mm.1 | TC0800000177.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3162604 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3162604 | - | - |
| mouse-6 v1.1 (Illumina) | GI_85702267-I | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3089574 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3162604 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3089574 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3162604 | - | - |

Spag11b
Genomic Map